Anti-DDK Tag Monoclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: OriGene
ORIGTA150030EA 597 EUR
ORIGTA150030
Anti-DDK Tag Monoclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Tag
Antigen: DDK Tag
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: None
Isotype: none
Reactivity:
Order Now