Anti-ZNF24 Mouse Monoclonal Antibody
Proveedor: Abgent
ABGEAT4613AEA 350 EUR
ABGEAT4613A
Anti-ZNF24 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: ZNF24 (zinc finger protein 24)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human