Anti-ZFP36L1 Mouse Monoclonal Antibody
Proveedor: Abgent
ABGEAT4574AEA 350 EUR
ABGEAT4574A
Anti-ZFP36L1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: ZFP36L1 (ZFP36 ring finger protein-like 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human