Anti-YWHAB Mouse Monoclonal Antibody

ABGEAT4558AEA 315 EUR
ABGEAT4558A
Anti-YWHAB Mouse Monoclonal Antibody
Type: Primary
Antigen: YWHAB (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR