Anti-UBE2M Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT4440AEA 350 EUR
ABGEAT4440A
Anti-UBE2M Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: UBE2M (ubiquitin-conjugating enzyme E2M)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR