Anti-TRIM49 Mouse Monoclonal Antibody
Proveedor: Abgent
ABGEAT4357AEA 350 EUR
ABGEAT4357A
Anti-TRIM49 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TRIM49 (tripartite motif containing 49)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human