Anti-TOP2A Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT4305AEA 350 EUR
ABGEAT4305A
Anti-TOP2A Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TOP2A (topoisomerase (DNA) II alpha 170kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR