Anti-TNFRSF21 Mouse Monoclonal Antibody
Proveedor: Abgent
ABGEAT4279AEA 350 EUR
ABGEAT4279A
Anti-TNFRSF21 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TNFRSF21 (tumor necrosis factor receptor superfamily, member 21)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human