Anti-TCF7L2 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT4190AEA 350 EUR
ABGEAT4190A
Anti-TCF7L2 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TCF7L2 (transcription factor 7-like 2 (T-cell specific, HMG-box))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR