Anti-TAF1A Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT4143AEA 350 EUR
ABGEAT4143A
Anti-TAF1A Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TAF1A (TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR