Anti-TAF1 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT4136AEA 350 EUR
ABGEAT4136A
Anti-TAF1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TAF1 (TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR