Anti-STAT3 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT4062AEA 350 EUR
ABGEAT4062A
Anti-STAT3 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR