Anti-SRF Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT4037AEA 350 EUR
ABGEAT4037A
Anti-SRF Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: SRF (serum response factor (c-fos serum response element-binding transcription factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR