Anti-SPON1 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT4027AEA 350 EUR
ABGEAT4027A
Anti-SPON1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: SPON1 (spondin 1, extracellular matrix protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR