Anti-SLC25A11 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT3910AEA 350 EUR
ABGEAT3910A
Anti-SLC25A11 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: SLC25A11 (solute carrier family 25 (mitochondrial carrier; oxoglutarate carrier), member 11)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR