Anti-SH3GLB1 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT3875AEA 350 EUR
ABGEAT3875A
Anti-SH3GLB1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: SH3GLB1 (SH3-domain GRB2-like endophilin B1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR