Anti-SERPINC1 Mouse Monoclonal Antibody
Proveedor: Abgent
ABGEAT3823AEA 350 EUR
ABGEAT3823A
Anti-SERPINC1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: SERPINC1 (serpin peptidase inhibitor, clade C (antithrombin), member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human