Anti-WDR61 Mouse Monoclonal Antibody
Proveedor: Abgent
ABGEAT3613AEA 350 EUR
ABGEAT3613A
Anti-WDR61 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: WDR61 (WD repeat domain 61)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human