Anti-RBPJ Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT3603AEA 350 EUR
ABGEAT3603A
Anti-RBPJ Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: RBPJ (recombination signal binding protein for immunoglobulin kappa J region)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR