Anti-RBP4 Mouse Monoclonal Antibody
Proveedor: Abgent
ABGEAT3601AEA 350 EUR
ABGEAT3601A
Anti-RBP4 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: RBP4 (retinol binding protein 4, plasma)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human