Anti-PRPF19 Mouse Monoclonal Antibody
Proveedor: Abgent
ABGEAT3441AEA 350 EUR
ABGEAT3441A
Anti-PRPF19 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: PRPF19 (pre-mRNA processing factor 19)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human