Anti-PRKAB2 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT3429AEA 350 EUR
ABGEAT3429A
Anti-PRKAB2 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: PRKAB2 (protein kinase, AMP-activated, beta 2 non-catalytic subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR