Anti-POMGNT1 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT3382AEA 350 EUR
ABGEAT3382A
Anti-POMGNT1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: POMGNT1 (protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR