Anti-PGGT1B Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT3281AEA 350 EUR
ABGEAT3281A
Anti-PGGT1B Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: PGGT1B (protein geranylgeranyltransferase type I, beta subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR