Anti-MYL6 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT2956AEA 350 EUR
ABGEAT2956A
Anti-MYL6 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: MYL6 (myosin, light chain 6, alkali, smooth muscle and non-muscle)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR