Anti-MS4A12 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT2908AEA 350 EUR
ABGEAT2908A
Anti-MS4A12 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: MS4A12 (membrane-spanning 4-domains, subfamily A, member 12)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR