Anti-MPP1 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT2893AEA 350 EUR
ABGEAT2893A
Anti-MPP1 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: MPP1 (membrane protein, palmitoylated 1, 55kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR