Anti-MPHOSPH10 Mouse Monoclonal Antibody

Proveedor: Abgent
ABGEAT2891AEA 350 EUR
ABGEAT2891A
Anti-MPHOSPH10 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: MPHOSPH10 (M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR