Water quality monitoring system, W-23XD series
Proveedor: Horiba
HORI3014007930EA 0 EUR
HORI3014007930
Water quality monitoring system, W-23XD series
Sistemas para monitorización Sistemas para monitorización y test de agua
W-23XD is capable of simultaneous measurements of as many as 13 parameters, including pH (option), DO (option), conductivity, salinity, TDS, seawater specific gravity, temperature, turbidity, depth, and ORP (option). The W-23XD also supports optional ion sensors and is capable of measurement at depths up to 100 metres. Sensor electronics are housed in a 47 mm probe and are suitable for operation in pressurised conditions.
  • Simultaneous monitoring of up to 13 parameters
  • Use in a wide range of salt or fresh water applications
  • Sensor probe has memory for up to one month of data at 15 minute intervals
  • Deploy sensor probe for measurement without control unit or cables
  • Compatible with GPS and data retrieval options
  • Manage retrieved data through PC
The probes have enough memory to store data from up to one month of continuous monitoring at 15 minute intervals. Because memory is stored in the sensor probe, it can be deployed without cables and the control unit for independent monitoring. The W-23XD sensor probe includes a depth sensor, conductivity-temperature-turbidity sensors, optional replaceable DO and pH/ORP sensors, and support for replaceable ion sensors. Optional sensors for use with the W-23XD include nitrate ion, calcium ion, fluoride ion, potassium ion, and ammonia. Cables are available in 2, 10, 30, 60, and 100 meters. Additional probes for other applications are available.
Información de suministro: System comprises probe with built-in sensors ( depth, conductivity, salinity, TDS, specific gravity [sea water], temperature and turbidity plus choice pH or ORP sensor plus choice of one ion sensor) and pH 4 standard solution, reference solution, Ion auto-cal solution, calibration beaker, batteries and instruction manual. Control unit and cables to be ordered separately.