Hand disinfectants, PURELL® Advanced hygienic hand rub
Proveedor: GOJO
PURELL® Advanced
115-1744EA 52.1 EUR
115-1744 GOJO9657-12-EEUOO 115-3461
Hand disinfectants, PURELL® Advanced hygienic hand rub
Limpieza de la piel Desinfectantes de la piel Desinfectantes para la piel líquidos y en espuma
A rub that feels great to use, with exceptional antimicrobial efficacy.
  • Bactericidal and virucidal
  • Clinically proven to maintain skin health
  • Refill bottles made from durable, recyclable PET, 30% less material than rigid HDPE (GOJO SMART-FLEX™)
  • Refill bottles snap into place quickly and easily (GOJO SANITARY SEALED™)
Colour: Clear
Certificaciones: EN 1500 (hand sanitiser), EN 12791 (surgical rub), EN 14476
Precaución: Use biocides safely. Always read the label and product information before use.