Fluorescent ruler set
Proveedor: Diversified Biotech
DIVEFRK-100EA 125 EUR
DIVEFRK-100
Fluorescent ruler set
Reglas
Fluorescent ruler set used to measure the mobility of electrophoretic markers and ethidium bromide-DNA complexes during electrophoresis.
  • Fluorescent ink markings: Sharp and photograph well under UV light (300 and 350 nm)
  • Single lane of agarose gel at 7 mm
  • Rulers made from flexible UV-clear plastic
  • Only the fluorescent markings will show under UV light
Información para pedidos: Includes two 10 cm rulers and two 20 cm rulers, 7 mm width.