Large cryogenic containers, BF series
478-0653EA 18500 EUR
VWRI478-0653 VWRI478-0645 VWRI478-0646 VWRI478-0649
Large cryogenic containers, BF series
The BF Series of cryogenic storage freezers offer a very high storage capacity for all types of biological samples. They are designed with high efficiency multilayer super-insulation and provide a reliable and efficient cryopreservation for samples stored in straws, cryovials or blood bags. Several features of design improve the ergonomics and bio-security for the sample and safety of the operator: large neck opening to offer easy access to all samples, the S170 CryoController for automatic filling, temperature and liquid nitrogen level measurement, alarm status and a data transfer supervision management system (optional), gas spring assisted lid opening with locking device, (except BF 2110).
  • Large neck opening of the container offers easy access to all samples
  • S170 CryoController for automatic filling of cryo-container, temperature and liquid nitrogen level measurement, alarm transmission, …
  • Data transfer towards a supervision management system (optional)
  • Gas spring assisted opening lid with locking device (except BF2110)
  • Protective cover on the lid insulation allows easy cleaning to avoid pollution
  • Optional rotating basket on BF2350PM and BF2600PM.
  • Possibility of storing in liquid, gas or dry phase
The units are offered with a wide range of polycarbonate, aluminium and stainless steel racks and all BF series can be used for storage of samples in liquid, gas or dry phase.
Protective cover on the lid to allow easy cleaning and avoid contamination, with the option of a rotating platform on BF 2350 PM and BF 2600 PM.
Certificaciones: The vessels are made in France with stainless steel, according to the highest quality levels.
The BF Series is compliant with European Medical Device Directive 93/42/EEC.