Sporicidal disinfectant, surface cleanser, Anioxy Spray WS

Proveedor: Anios
DMDA1756573EA 0 EUR
DMDA1756573
Sporicidal disinfectant, surface cleanser, Anioxy Spray WS
Soluciones de limpieza Desinfectantes
A ready to use disinfectant spray with broad spectrum activity for rapid cleaning of surfaces in the healthcare industry. It is not suitable for medical devices which require submersive cleaning eg endoscopes.

  • Bactericidal, tuberculocide, fungicidal (yeast), sporicidal, active on bacillus subtilis spores, clostridium difficile spores, active on GRP, BVDV, rotavirus and PRV virus
  • Contains hydrogen peroxide and ethanol in an aqueous medium
  • Leaves no residue after use
  • pH neutral and chlorine-free

Certificaciones: Bactericide EN 1040, EN 13727; Tuberculocide EN 14563; Fungicide EN 1275, EN 13624; Sporicidal EN 13704
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR