Mop cover CleanPad
BASA130876EA 0 EUR
BASA130876
Mop cover CleanPad
CleanPad mop covers are constructed from 100% single-knit continuous polyester filament. Due to a simple elasticated system, they can easily be installed and replaced over the CleanPad mop head (20×38 cm). Ultra clean and absorbent, the CleanPad mop covers are the best solution to clean walls, floors and ceilings in all cleanroom environments. In conjunction with the included foam pad, CleanPad mop covers ensure an efficient cleaning even on textured surfaces (the foam pads can also be ordered separately, allowing frequent replacement).