Cannon-Fenske viscometers, UKAS
Proveedor: Fungilab
FUNGCV004-202EA 93.73 EUR
FUNGCV004-202 FUNGCV004-203 FUNGCV004-204 FUNGCV004-205 FUNGCV004-206 FUNGCV004-207 FUNGCV004-208 FUNGCV004-209 FUNGCV004-210 FUNGCV004-211 FUNGCV004-212 FUNGCV004-201
Cannon-Fenske viscometers, UKAS
Viscosímetros Viscosímetros capilares
Cannon-Fenske viscometers can measure the viscosities of transparent Newtonian liquids.
  • Constant value indication (±0,2%) at 40 and 100 °C
Suitable for applications in the chemical and petroleum industries.
Certificaciones: UNE 400313, ASTM D-445-446, ASTM D2515 and ISO 3104-3105.
Información de suministro: Supplied with calibration certificate.