Rotary vane vacuum pumps, one-stages, type PS
Proveedor: Gardner Denver
ILMV321021EA 0 EUR
ILMV321021 ILMV321020
Rotary vane vacuum pumps, one-stages, type PS
Bombas Bombas de paletas giratorias de vacío
These rotary vane vacuum pumps have high pumping speed that makes them ideal to be used in laboratory or industrial applications. Due to their high water vapor tolerance and outstanding reliability, these pumps are perfect for chemical applications.
  • Compact design (small footprint and low weight)
  • Low sound level on daily operations
Pumps require low maintenance which can be handled quickly and simply with predefined service kits. To prevent accidental damage, the oil sight glass is integrated into the housing. Due to the compact design, its easy for wipe cleaning.
Used widely in vacuum oven, industrial, drying and degassing.
Información de suministro: Supplied with an anti-suck back valve (P-E series), oil charging, gas ballast valve, centering ring, clamping ring and motor overload protection.