Direct thermal Cryo-Tags® labels
Proveedor: Diversified Biotech
Cryo-Tags®
DIVEDTCR-3000EA 156 EUR
DIVEDTCR-3000 DIVEDTCR-4000 DIVEDTCR-5000 DIVEDTCR-6000 DIVEDTCR-1000 DIVEDTCR-2000
Direct thermal Cryo-Tags® labels
Etiquetas
These Cryo-Tags® polypropylene cryo labels accept printing from direct thermal printers such as Dymo® LabelWriter® to print popular barcode formats for fast easy sample identification.
  • Temperature resistance from −196 to +70 °C
  • Freezable in liquid and vapour phase nitrogen
  • Adhere to most plastics, glass and metals without cracking, peeling or degrading, scuffing or smearing
Tubes labeled with direct thermal Cryo-Tags® easily slide in and out of centrifuge rotors without binding.
Precaución: Only compatible with US model Dymo® labelWriters®.