Transport swab with medium
Proveedor: Deltalab
DELT300297EA 101 EUR
DELT300297
Transport swab with medium
Hisopos Hisopos para transporte
This viral transport medium preserves the specimen during transport to the laboratory.
  • Sterilised by irradiaton
  • Medium to maintain the sample viable for 72 h
  • Antimicrobial substances inhibit the growth of bacteria and fungi
Suitable for nasal, pharyngeal, ocular, skin and mucus samples.
Precaución: * Los productos de esta página están fuera del alcance de la Directiva de la UE 2019/904