4-(Diethylamino)azobenzene ≥97.0% (by HPLC, titration analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAD1126-100GEA
116
EUR
TCIAD1126-100G
4-(Diethylamino)azobenzene ≥97.0% (by HPLC, titration analysis)
4-(Diethylamino)azobenzene
Formula:
C₁₆H₁₉N₃ MW: 253,35 g/mol |
Número MDL:
MFCD00009043 Núm. CAS: 2481-94-9 EINECS: 219-616-8 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 95.0 to 98.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...