2-(2-Chlorophenoxy)propionic acid ≥99.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAC1096-25GEA
138
EUR
TCIAC1096-25G
2-(2-Chlorophenoxy)propionic acid ≥99.0% (by titrimetric analysis)
2-(2-Chlorophenoxy)propionic acid
Formula:
C₉H₉ClO₃ |
Número MDL:
MFCD00002644 Núm. CAS: 25140-86-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 114.0 to 117.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...