4,4'-Methylenebis(2-chloroaniline), Supelco®


M-Clarity™ quality level = MQ100

66681-1GEA 67.3 EUR
66681-1G 66681-250MG
4,4'-Methylenebis(2-chloroaniline), Supelco®
4,4'-Methylenebis(2-chloroaniline)
Formula: C₁₃H₁₂Cl₂N₂
Número MDL: MFCD00047829
Núm. CAS: 101-14-4
EINECS: 202-918-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR