N-Boc-L-phenylalaninol ≥97.0% (por GC)
Proveedor: TCI EUROPE
TCIAB2604-5GEA
48
EUR
TCIAB2604-5G
N-Boc-L-phenylalaninol ≥97.0% (por GC)
N-Boc-L-phenylalaninol
Formula:
C₁₄H₂₁NO₃ MW: 251,3 g/mol |
Número MDL:
MFCD00076976 Núm. CAS: 66605-57-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 94.0 to 98.0 deg-C |
Specific rotation [a]20/D | -25.0 to -30.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...