1,5-Bis(4-aminophenoxy)pentane ≥97%

Proveedor: Thermo Scientific

B21434.06EA 120 EUR
B21434.06
1,5-Bis(4-aminophenoxy)pentane ≥97%
1,5-Bis(4-aminophenoxy)pentane
Formula: C₁₇H₂₂N₂O₂
Número MDL: MFCD01632139
Núm. CAS: 2391-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR