1,3-Propanedisulphonic acid disodium salt ≥97.0% (by titrimetric analysis)
Proveedor: TCI EUROPE
|
Atención
|
TCIAP1205-5GEA
30
EUR
TCIAP1205-5G
1,3-Propanedisulphonic acid disodium salt ≥97.0% (by titrimetric analysis)
1,3-Propanedisulphonic acid disodium salt
Formula:
C₃H₆Na₂O₆S₂ MW: 248,19 g/mol |
Número MDL:
MFCD00191483 Núm. CAS: 36589-58-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...