Perfluoro(2-butyltetrahydrofuran) (so called)
Proveedor: TCI EUROPE
TCIAP0867-25GEA
54
EUR
TCIAP0867-25G
Perfluoro(2-butyltetrahydrofuran) (so called)
Perfluoro(2-butyltetrahydrofuran)
Formula:
C₈F₁₆O MW: 416,06 g/mol Pto. Ebullición: 102°C |
Número MDL:
MFCD00014513 Núm. CAS: 335-36-4 EINECS: 206-389-5 |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | multi component mixture |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...