Dimetridazole ≥97.0% (by titrimetric analysis) for biochemical research
Proveedor: TCI EUROPE
Atención
TCIAD4081-5GEA 21 EUR
TCIAD4081-5G TCIAD4081-25G
Dimetridazole ≥97.0% (by titrimetric analysis) for biochemical research
Dimetridazole
Formula: C₅H₇N₃O₂
MW: 141,13 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00047046
Núm. CAS: 551-92-8
EINECS: 209-001-2
https://es.vwr.com/stibo/web/std.lang.all/60/29/28986029.jpg
Order Now
Resultados test especificaciones
Appearance White to Yellow to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 137.0 to 141.0 deg-C
Protein impurity test (SDS-PAGE) to pass test