Dimetridazole
Proveedor: EHRENSTORFER GMBH
EHERC12772000EA
87.5
EUR
EHERC12772000
Dimetridazole
Dimetridazole
Formula:
C₅H₇N₃O₂ MW: 141,13 g/mol |
Número MDL:
MFCD00047046 Núm. CAS: 551-92-8 EINECS: 209-001-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...