5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate
Proveedor: Hach
HACH230424EA
195
EUR
HACH230424
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate
Formula:
C₂₀H₁₃N₄NaO₆S₂·H₂O MW: 492,47 g/mol (anhy) |
Número MDL:
MFCD00150794 Núm. CAS: 69898-45-9 EINECS: 274-196-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...