2-(2-Naphthyl)indole ≥98.0% (por GC)
Proveedor: TCI EUROPE
TCIAN0859-1GEA
84
EUR
TCIAN0859-1G
2-(2-Naphthyl)indole ≥98.0% (por GC)
2-(2-Naphthyl)indole
Formula:
C₁₈H₁₃N MW: 243,31 g/mol |
Número MDL:
MFCD00124144 Núm. CAS: 23746-81-8 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 202.0 to 206.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...