Pilocarpine nitrate
EDQMP1700000EA
235
EUR
EDQMP1700000
Pilocarpine nitrate
Pilocarpine nitrate
Formula:
C₁₁H₁₇N₃O₅ |
Número MDL:
MFCD00078497 Núm. CAS: 148-72-1 EINECS: 205-723-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...